missing translation for 'onlineSavingsMsg'
Learn More

4-Biphenylacetonitrile, 97%, Thermo Scientific Chemicals

Product Code. 11364116
Change view
Click to view available options
Quantity:
500 mg
2 g
Unit Size:
2g
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11364116 2 g 2g
11354116 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11364116 Supplier Thermo Scientific Alfa Aesar Supplier No. L19243.04
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 31603-77-7 | Molecular Formula: C14H11N | Molecular Weight: 193.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31603-77-7
Molecular Formula C14H11N
Molecular Weight (g/mol) 193.25
MDL Number MFCD00016403
InChI Key HSZCNGTZJWZAMF-UHFFFAOYSA-N
Synonym 4-biphenylacetonitrile, 2-1,1'-biphenyl-4-yl acetonitrile, 2-4-phenylphenyl acetonitrile, 1,1'-biphenyl-4-acetonitrile, 4-cyanomethylbiphenyl, p-biphenylacetonitrile, p-biphenylylacetonitrile, 4-biphenyl-acetonitrile, 4-biphenylylacetonitrile, p-phenyl benzyl cyanide
PubChem CID 35856
IUPAC Name 2-(4-phenylphenyl)acetonitrile
SMILES N#CCC1=CC=C(C=C1)C1=CC=CC=C1

Specifications

Melting Point 93°C to 97°C
Boiling Point 146°C to 150°C (0.03 mmHg)
Quantity 2 g
Beilstein 1867255
Formula Weight 193.25
Percent Purity 97%
Chemical Name or Material 4-Biphenylacetonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.