missing translation for 'onlineSavingsMsg'
Learn More

4-Biphenylacetic acid, 98%

Product Code. 10175583 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
500 g
Packaging:
Glass Bottle
Unit Size:
100g
10g
25g
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10175583 500 g Glass Bottle 500g
10578873 10 g Glass Bottle 10g
10367700 25 g Glass Bottle 25g
10696012 100 g Glass Bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10175583 Supplier Thermo Scientific Acros Supplier No. 188155000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 5728-52-9 | C14H12O2 | 212.248 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5728-52-9
MDL Number MFCD00004351
InChI Key QRZAKQDHEVVFRX-UHFFFAOYSA-N
Synonym 4-biphenylacetic acid, felbinac, 4-biphenylylacetic acid, traxam, dolinac, 4-carboxymethylbiphenyl, napageln, 2-4-phenylphenyl acetic acid, p-biphenylylacetic acid, 1,1'-biphenyl-4-acetic acid
PubChem CID 3332
ChEBI CHEBI:31597
IUPAC Name 2-(4-phenylphenyl)acetic acid
SMILES C1=CC=C(C=C1)C2=CC=C(C=C2)CC(=O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 156°C to 160°C
Color Beige
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Molecular Formula C14H12O2
Linear Formula C6H5C6H4CH2CO2H
Quantity 500 g
Beilstein 09, I, 284
Merck Index 15, 3985
Molecular Weight (g/mol) 212.25
Formula Weight 212.25
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 4-Biphenylacetic acid
Show More Show Less
compliance-icons
Product Identifier
  • 4-Biphenylacetic acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.