missing translation for 'onlineSavingsMsg'
Learn More

4-Biphenylacetic acid, 98%

Product Code. 11449293
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11449293 50 g 50g
11439293 10 g 10g
11459293 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11449293 Supplier Thermo Scientific Alfa Aesar Supplier No. A11749.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 5728-52-9 | C14H12O2 | 212.248 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5728-52-9
Molecular Formula C14H12O2
Molecular Weight (g/mol) 212.248
MDL Number MFCD00004351
InChI Key QRZAKQDHEVVFRX-UHFFFAOYSA-N
Synonym 4-biphenylacetic acid, felbinac, 4-biphenylylacetic acid, traxam, dolinac, 4-carboxymethylbiphenyl, napageln, 2-4-phenylphenyl acetic acid, p-biphenylylacetic acid, 1,1'-biphenyl-4-acetic acid
PubChem CID 3332
ChEBI CHEBI:31597
IUPAC Name 2-(4-phenylphenyl)acetic acid
SMILES C1=CC=C(C=C1)C2=CC=C(C=C2)CC(=O)O

Specifications

Melting Point 158°C to 163°C
Odor Odorless
Quantity 50 g
UN Number UN2811
Beilstein 1211592
Merck Index 14,3951
Formula Weight 212.25
Percent Purity 98%
Chemical Name or Material 4-Biphenylacetic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.