missing translation for 'onlineSavingsMsg'
Learn More

4-Benzylpyridine, 97%, Thermo Scientific™

Product Code. 10665902 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 mL
100 mL
Packaging:
Glass bottle
Unit Size:
100mL
5mL
This item is not returnable. View return policy

Product Code. 10665902

Brand: Thermo Scientific Acros 106161000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 2116-65-6
Molecular Formula C12H11N
Molecular Weight (g/mol) 169.23
MDL Number MFCD00006443
InChI Key DBOLXXRVIFGDTI-UHFFFAOYSA-N
Synonym pyridine, 4-phenylmethyl, pyridine, 4-benzyl, 4-benzyl-pyridine, gamma-benzylpyridine, .gamma.-benzylpyridine, 4-phenylmethyl pyridine, unii-1p29gqm0oh, 1p29gqm0oh, 4-benzyl pyridine, 4-benzylpyridine
PubChem CID 16458
IUPAC Name 4-benzylpyridine
SMILES C(C1=CC=CC=C1)C1=CC=NC=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 9.0°C to 11.0°C
Color Yellow
Density 1.0600g/mL
Boiling Point 287.0°C
Flash Point 115°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula C6H5CH2(C5H4N)
Refractive Index 1.5810 to 1.5830
Quantity 100 mL
Beilstein 20, 426
Specific Gravity 1.06
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in organic solvents,soluble in ethanol and ether
Formula Weight 169.23
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 4-Benzylpyridine, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Benzylpyridine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.