missing translation for 'onlineSavingsMsg'
Learn More

4-Benzylpyridine, 97+%

Product Code. 11309429
Change view
Click to view available options
Quantity:
50 g
100 g
500 g
Unit Size:
100g
500g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11309429 100 g 100g
11466877 50 g 50g
11319429 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11309429 Supplier Thermo Scientific Alfa Aesar Supplier No. A14326.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2116-65-6 | C12H11N | 169.23 g/mol

4-Benzylpyridine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, dyes and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Benzylpyridine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, dyes and agrochemicals.

Solubility
Soluble in ethanol, ethyl ether. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Incompatible with strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2116-65-6
Molecular Formula C12H11N
Molecular Weight (g/mol) 169.23
MDL Number MFCD00006443
InChI Key DBOLXXRVIFGDTI-UHFFFAOYSA-N
Synonym pyridine, 4-phenylmethyl, pyridine, 4-benzyl, 4-benzyl-pyridine, gamma-benzylpyridine, .gamma.-benzylpyridine, 4-phenylmethyl pyridine, unii-1p29gqm0oh, 1p29gqm0oh, 4-benzyl pyridine, 4-benzylpyridine
PubChem CID 16458
IUPAC Name 4-benzylpyridine
SMILES C(C1=CC=CC=C1)C1=CC=NC=C1

Specifications

Melting Point 9°C to 11°C
Density 1.061
Boiling Point 286°C to 287°C
Flash Point 115°C (239°F)
Refractive Index 1.5805
Quantity 100 g
Beilstein 115831
Solubility Information Soluble in ethanol,ethyl ether. Insoluble in water.
Formula Weight 169.23
Percent Purity 99%
Chemical Name or Material 4-Benzylpyridine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.