missing translation for 'onlineSavingsMsg'
Learn More

4-Benzylpiperidine, 98%

Product Code. 11496726
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11496726 25 g 25g
11406736 100 g 100g
11416736 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11496726 Supplier Thermo Scientific Alfa Aesar Supplier No. B23883.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 31252-42-3 | C12H17N | 175.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31252-42-3
Molecular Formula C12H17N
Molecular Weight (g/mol) 175.28
MDL Number MFCD00006006
InChI Key ABGXADJDTPFFSZ-UHFFFAOYSA-N
Synonym piperidine, 4-phenylmethyl, 4-phenylmethyl piperidine, 4-benzyl-piperidine, piperidine, 4-benzyl, phenyl 4-piperidyl methane, alpha-4-piperidyl toluene, unii-jdf1t4667s, 4-benzyl piperidine, 4-benzylpiperdine, 4-benzylpiperidin
PubChem CID 31738
IUPAC Name 4-benzylpiperidine
SMILES C(C1CCNCC1)C1=CC=CC=C1

Specifications

Melting Point 6°C to 7°C
Density 0.997
Boiling Point 278°C to 279°C
Flash Point 147°C (296°F)
Refractive Index 1.537
Quantity 25 g
Beilstein 132339
Formula Weight 175.28
Percent Purity 98%
Chemical Name or Material 4-Benzylpiperidine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.