missing translation for 'onlineSavingsMsg'
Learn More

4-Benzylpiperidine, 98%, Thermo Scientific Chemicals

Catalog Number p-65570
Click to view available options
Quantity:
5 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
31252-42-3
C12H17N
175.28
MFCD00006006
ABGXADJDTPFFSZ-UHFFFAOYSA-N
piperidine, 4-phenylmethyl, 4-phenylmethyl piperidine, 4-benzyl-piperidine, piperidine, 4-benzyl, phenyl 4-piperidyl methane, alpha-4-piperidyl toluene, unii-jdf1t4667s, 4-benzyl piperidine, 4-benzylpiperdine, 4-benzylpiperidin
31738
4-benzylpiperidine
C(C1CCNCC1)C1=CC=CC=C1
This item is not returnable. View return policy
31252-42-3
C12H17N
175.28
MFCD00006006
ABGXADJDTPFFSZ-UHFFFAOYSA-N
piperidine, 4-phenylmethyl, 4-phenylmethyl piperidine, 4-benzyl-piperidine, piperidine, 4-benzyl, phenyl 4-piperidyl methane, alpha-4-piperidyl toluene, unii-jdf1t4667s, 4-benzyl piperidine, 4-benzylpiperdine, 4-benzylpiperidin
31738
4-benzylpiperidine
C(C1CCNCC1)C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 6.0°C to 7.0°C
Color Colorless to Yellow
Density 0.9900g/mL
Boiling Point 279.0°C
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Refractive Index 1.5370 to 1.5390
Quantity 5 g
Beilstein 20, 296
Specific Gravity 0.99
Formula Weight 175.27
Percent Purity 98%
Physical Form Viscous Liquid
Chemical Name or Material 4-Benzylpiperidine, 98%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'