missing translation for 'onlineSavingsMsg'
Learn More

4-Benzyloxyphenylacetic Acid, TRC

Product Code. 30334061
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30334061 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30334061 Supplier Toronto Research Chemicals Supplier No. TRCB2877755G
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Benzyloxyphenylacetic Acid
CAS 6547-53-1
Molecular Formula C15H14O3
Quantity 5 g
Synonym 2-(4-Benzyloxyphenyl)acetic Acid, NSC 134561, p-Benzyloxyphenylacetic Acid
InChI Formula InChI=1S/C15H14O3/c16-15(17)10-12-6-8-14(9-7-12)18-11-13-4-2-1-3-5-13/h1-9H,10-11H2,(H,16,17)
SMILES OC(=O)Cc1ccc(OCc2ccccc2)cc1
IUPAC Name 2-(4-phenylmethoxyphenyl)acetic acid
Molecular Weight (g/mol) 242.27
Formula Weight 242.0943
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.