missing translation for 'onlineSavingsMsg'
Learn More

(4-Benzyloxyphenyl)acetic acid, 99%, Thermo Scientific™

Product Code. 10369563 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10369563 5 g Glass bottle 5g
10186163 1 g Glass bottle 1g
10513944 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10369563 Supplier Thermo Scientific Acros Supplier No. 394080050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 6547-53-1
Molecular Formula C15H14O3
Molecular Weight (g/mol) 242.27
MDL Number MFCD00017540
InChI Key XJHGAJLIKDAOPE-UHFFFAOYSA-N
Synonym 4-benzyloxyphenylacetic acid, 2-4-benzyloxy phenyl acetic acid, 4-benzyloxy phenyl acetic acid, 4-benzyloxyphenyl acetic acid, benzeneacetic acid, 4-phenylmethoxy, 4-benzyloxy phenylacetic acid, 4-benzyloxy-phenyl-acetic acid, 2-4-phenylmethoxyphenyl acetic acid, 2-4-phenylmethoxy phenyl acetic acid
PubChem CID 81033
IUPAC Name 2-(4-phenylmethoxyphenyl)acetic acid
SMILES C1=CC=C(C=C1)COC2=CC=C(C=C2)CC(=O)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 120°C to 122°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Linear Formula C6H5CH2OC6H4CH2CO2H
Quantity 5 g
Formula Weight 242.27
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material (4-Benzyloxyphenyl)acetic acid
Show More Show Less
compliance-icons
Product Identifier
  • (4-Benzyloxyphenyl)acetic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.