missing translation for 'onlineSavingsMsg'
Learn More

4-Benzyloxyphenol, 98+%

Product Code. p-7022196
Click to view available options
Quantity:
25 g
50 g
100 g
250 g
1000 g
Unit Size:
1000g
100g
250g
25g
50g
This item is not returnable. View return policy

Product Code. 11329429

Brand: Thermo Scientific Alfa Aesar A14843.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 103-16-2 | C13H12O2 | 200.237 g/mol

4-Benzyloxyphenol is used in the synthesis of bis(4-benzyloxyphenoxy)phenyl phosphine oxide. It plays an essential role in the preparation of hetaryl-azophenol dyes via heterocyclic amines in nitrosyl sulphuric acid. It is also used for polyester fiber dyeing and in rubber industry. It acts as a depigmenting agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Benzyloxyphenol is used in the synthesis of bis(4-benzyloxyphenoxy)phenyl phosphine oxide. It plays an essential role in the preparation of hetaryl-azophenol dyes via heterocyclic amines in nitrosyl sulphuric acid. It is also used for polyester fiber dyeing and in rubber industry. It acts as a depigmenting agent.

Solubility
Slightly soluble in water. Soluble in alcohol, acetone, benzene, and diethyl ether.

Notes
Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103-16-2
Molecular Formula C13H12O2
MDL Number MFCD00002333
InChI Key VYQNWZOUAUKGHI-UHFFFAOYSA-N
Synonym 4-benzyloxyphenol, monobenzone, 4-benzyloxy phenol, hydroquinone monobenzyl ether, benoquin, benzoquin, superlite, monobenzyl hydroquinone, leucodinine, monobenzon
PubChem CID 7638
ChEBI CHEBI:34380
IUPAC Name 4-phenylmethoxyphenol

Specifications

Melting Point 118°C to 122°C
Density 1.26
Quantity 25 g
Beilstein 1958305
Merck Index 14,6248
Solubility Information Slightly soluble in water. Soluble in alcohol,acetone,benzene,and diethyl ether.
SMILES C1=CC=C(C=C1)COC2=CC=C(C=C2)O
Molecular Weight (g/mol) 200.237
Formula Weight 200.24
Percent Purity ≥98%
Chemical Name or Material 4-Benzyloxyphenol
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.