missing translation for 'onlineSavingsMsg'
Learn More

4-Benzyloxybenzyl alcohol, 99%

Product Code. 10576423 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
10g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10576423 10 g Glass bottle 10g
10512211 25 g Glass bottle 25g
10157663 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10576423 Supplier Thermo Scientific Acros Supplier No. 181960100
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 836-43-1 | C14H14O2 | 214.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 836-43-1
Molecular Formula C14H14O2
Molecular Weight (g/mol) 214.26
MDL Number MFCD00004654
InChI Key OEBIVOHKFYSBPE-UHFFFAOYSA-N
Synonym 4-benzyloxybenzyl alcohol, 4-benzyloxy phenyl methanol, p-benzyloxybenzyl alcohol, 4-benzyloxybenzylalcohol, 4-benzyloxy benzyl alcohol, benzenemethanol, 4-phenylmethoxy, p-alkoxybenzyl alcohol resin, 4-phenylmethoxyphenyl methanol, argopore-wang
PubChem CID 70043
ChEBI CHEBI:29486
SMILES OCC1=CC=C(OCC2=CC=CC=C2)C=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 84°C to 87°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula C6H5CH2OC6H4CH2OH
Quantity 10 g
IUPAC Name (4-phenylmethoxyphenyl)methanol
Formula Weight 214.26
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 4-Benzyloxybenzyl alcohol
Show More Show Less
compliance-icons
Product Identifier
  • 4-Benzyloxybenzyl alcohol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.