missing translation for 'onlineSavingsMsg'
Learn More

4-Benzyloxybenzyl alcohol, 98+%

Product Code. 11457137
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11457137 10 g 10g
11467137 50 g 50g
11477137 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11457137 Supplier Thermo Scientific Alfa Aesar Supplier No. A14949.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 836-43-1 | C14H14O2 | 214.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 836-43-1
Molecular Formula C14H14O2
Molecular Weight (g/mol) 214.26
MDL Number MFCD00004654
InChI Key OEBIVOHKFYSBPE-UHFFFAOYSA-N
Synonym 4-benzyloxybenzyl alcohol, 4-benzyloxy phenyl methanol, p-benzyloxybenzyl alcohol, 4-benzyloxybenzylalcohol, 4-benzyloxy benzyl alcohol, benzenemethanol, 4-phenylmethoxy, p-alkoxybenzyl alcohol resin, 4-phenylmethoxyphenyl methanol, argopore-wang
PubChem CID 70043
ChEBI CHEBI:29486
IUPAC Name (4-phenylmethoxyphenyl)methanol
SMILES OCC1=CC=C(OCC2=CC=CC=C2)C=C1

Specifications

Melting Point 84°C to 88°C
Quantity 10 g
Beilstein 1877819
Formula Weight 214.26
Percent Purity ≥98%
Chemical Name or Material 4-Benzyloxybenzyl alcohol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.