missing translation for 'onlineSavingsMsg'
Learn More

4-Benzyloxybenzotrifluoride, 95%, Thermo Scientific™

Product Code. 15407828
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 70097-65-3
Molecular Formula C14H11F3O
Molecular Weight (g/mol) 252.236
MDL Number MFCD00275905
InChI Key SNRFAICCYVUTRG-UHFFFAOYSA-N
Synonym 1-benzyloxy-4-trifluoromethyl benzene, 4-benzyloxybenzotrifluoride, 4-benzyloxy benzotrifluoride, 1-phenylmethoxy-4-trifluoromethyl benzene, maybridge1_001022, 4 benzyloxy benzotrifluoride, 4-trifluoromethylphenyl benzyl ether, 1-benzyloxy-4-trifluoromethylbenzene, 1-benzyloxy-4-trifluoromethyl-benzene
PubChem CID 2798168
IUPAC Name 1-phenylmethoxy-4-(trifluoromethyl)benzene
SMILES C1=CC=C(C=C1)COC2=CC=C(C=C2)C(F)(F)F

Specifications

Quantity 250 mg
Formula Weight 252.23
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 4-Benzyloxybenzotrifluoride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'