missing translation for 'onlineSavingsMsg'
Learn More

4-Benzyloxybenzoic acid, 98%

Product Code. 11437697
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11437697 25 g 25g
11417697 1 g 1g
11427697 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11437697 Supplier Thermo Scientific Alfa Aesar Supplier No. A15715.14
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 1486-51-7 | C14H12O3 | 228.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1486-51-7
Molecular Formula C14H12O3
Molecular Weight (g/mol) 228.25
MDL Number MFCD00016527
InChI Key AQSCHALQLXXKKC-UHFFFAOYSA-N
PubChem CID 73880
IUPAC Name 4-phenylmethoxybenzoic acid
SMILES OC(=O)C1=CC=C(OCC2=CC=CC=C2)C=C1

Specifications

Melting Point 187°C to 191°C
Quantity 25 g
Beilstein 2111640
Synonym 4-benzyloxybenzoic acid, 4-benzyloxy benzoic acid, benzoic acid, 4-phenylmethoxy, p-benzyloxy benzoic acid, 4-phenylmethoxy benzoic acid, 4-benzyloxy-benzoic acid, 4-benzyl-oxy benzoic acid, benzoic acid,4-phenylmethoxy, asischem a95731
Formula Weight 228.25
Percent Purity 98%
Chemical Name or Material 4-Benzyloxybenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.