missing translation for 'onlineSavingsMsg'
Learn More

4-Benzyloxybenzaldehyde, 98%

Product Code. 11488466
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11488466 25 g 25g
11498466 100 g 100g
11408476 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11488466 Supplier Thermo Scientific Alfa Aesar Supplier No. A15934.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 4397-53-9 | C14H12O2 | 212.25 g/mol

4-Benzyloxybenzaldehyde is used in the synthesis of (5-fluoro-(2R*,3S*)-2,3-bis(4-hydroxyphenyl)pentanenitrile),an estrogen receptor β-selective ligand. It is a position isomer of the adenylyl cyclase activator 2-benzyloxybenzaldehyde.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Benzyloxybenzaldehyde is used in the synthesis of (5-fluoro-(2R*,3S*)-2,3-bis(4-hydroxyphenyl)pentanenitrile),an estrogen receptor β-selective ligand. It is a position isomer of the adenylyl cyclase activator 2-benzyloxybenzaldehyde.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. Store under dry inert gas. It is sensitive to air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4397-53-9
Molecular Formula C14H12O2
Molecular Weight (g/mol) 212.25
MDL Number MFCD00003387,MFCD01632880
InChI Key ZVTWZSXLLMNMQC-UHFFFAOYSA-N
Synonym 4-benzyloxybenzaldehyde, 4-benzyloxy benzaldehyde, benzaldehyde, 4-phenylmethoxy, p-benzyloxy benzaldehyde, benzaldehyde, p-benzyloxy, 4-benzyloxy-benzaldehyde, 4-phenylmethoxy benzaldehyde, 4-benzyl-oxy benzaldehyde, wang aldehyde resin
PubChem CID 78109
IUPAC Name 4-phenylmethoxybenzaldehyde
SMILES O=CC1=CC=C(OCC2=CC=CC=C2)C=C1

Specifications

Melting Point 69°C to 74°C
Boiling Point 197°C to 199°C (20 mmHg)
Quantity 25 g
Beilstein 1242385
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 212.25
Percent Purity 98%
Chemical Name or Material 4-Benzyloxybenzaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.