missing translation for 'onlineSavingsMsg'
Learn More

4-(Benzyloxy)phenol, TRC

Product Code. 30345365
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30345365 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30345365 Supplier Toronto Research Chemicals Supplier No. TRCB28819025G
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-(Benzyloxy)phenol
CAS 103-16-2
Molecular Formula C13 H12 O2
Quantity 25 g
Synonym Phenol, p-(benzyloxy)- (6CI,7CI,8CI), 4-(Phenylmethoxy)phenol, 4-(Benzyloxy)phenol, AgeRite Alba, Agerite, Alba-Dome, Benoquin, Benzoquin, Benzyl p-hydroxyphenyl ether, Carmifal, Depigman, Dermochinona, Hydroquinone benzyl ether, Hydroquinone monobenzyl ether, Leucodinine, Monobenzon, Monobenzone, Monobenzyl ether hydroquinone, Monobenzyl hydroquinone, NSC 2132, NSC 33918, Pigmex, Superlite, Superlite (antioxidant), p-(Benzyloxy)phenol, p-Hydroxyphenyl benzyl ether
InChI Formula InChI=1S/C13H12O2/c14-12-6-8-13(9-7-12)15-10-11-4-2-1-3-5-11/h1-9,14H,10H2
SMILES Oc1ccc(OCc2ccccc2)cc1
IUPAC Name 4-phenylmethoxyphenol
Molecular Weight (g/mol) 200.23
Formula Weight 200.0837
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.