missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Benzyloxy-3-methoxybenzaldehyde, TRC
Click to view available options
Quantity:
10 g
Unit Size:
10g
Specifications
Specifications
| Chemical Name or Material | 4-Benzyloxy-3-methoxybenzaldehyde |
| CAS | 2426-87-1 |
| Molecular Formula | C15H14O3 |
| Quantity | 10 g |
| Synonym | 4-(Benzyloxy)-3-methoxy-benzaldehyde, 4-(Benzyloxy)-3-methoxybenzaldehyde, 4-O-Benzylvanillin, Benzylvanillin, NSC 208757, NSC 22599, NSC 44876, O-Benzylvanillin, Vanillin Benzyl Ether |
| InChI Formula | InChI=1S/C15H14O3/c1-17-15-9-13(10-16)7-8-14(15)18-11-12-5-3-2-4-6-12/h2-10H,11H2,1H3 |
| SMILES | COc1cc(C=O)ccc1OCc2ccccc2 |
| IUPAC Name | 3-methoxy-4-phenylmethoxybenzaldehyde |
| Molecular Weight (g/mol) | 242.27 |
| Formula Weight | 242.0943 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction