missing translation for 'onlineSavingsMsg'
Learn More

4-Benzyloxy-3-methoxybenzaldehyde, 98%

Product Code. 11497736
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11497736 25 g 25g
11407746 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11497736 Supplier Thermo Scientific Alfa Aesar Supplier No. A14716.14
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2426-87-1 | C15H14O3 | 242.27 g/mol

4-Benzyloxy-3-methoxybenzaldehyde was used in the synthesis of 1,2-bis(4-benzyloxy-3-methoxyphenyl)-3-hydroxy-propionic acid1. It was also used in first enantioselective total synthesis of a neurotrophic (-)-talaumidin. 4-Benzyloxy-3-methoxybenzaldehyde reacts with benzohydrazide to yield (E)-N?-(4-benzyloxy-3-methoxybenzylidene)benzohydrazide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Benzyloxy-3-methoxybenzaldehyde was used in the synthesis of 1,2-bis(4-benzyloxy-3-methoxyphenyl)-3-hydroxy-propionic acid1. It was also used in first enantioselective total synthesis of a neurotrophic (-)-talaumidin. 4-Benzyloxy-3-methoxybenzaldehyde reacts with benzohydrazide to yield (E)-N′-(4-benzyloxy-3-methoxybenzylidene)benzohydrazide.

Solubility
Soluble in chloroform, methanol.

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2426-87-1
Molecular Formula C15H14O3
Molecular Weight (g/mol) 242.27
MDL Number MFCD00003365
InChI Key JSHLOPGSDZTEGQ-UHFFFAOYSA-N
Synonym 4-benzyloxy-3-methoxybenzaldehyde, benzylvanillin, o-benzylvanillin, vanillin benzyl ether, unii-5knc2tsw9g, 4-benzyloxy-3-methoxy-benzaldehyde, benzaldehyde, 3-methoxy-4-phenylmethoxy, 5knc2tsw9g, 3-methoxy-4-phenylmethoxy benzaldehyde
PubChem CID 75506
IUPAC Name 3-methoxy-4-phenylmethoxybenzaldehyde
SMILES COC1=CC(C=O)=CC=C1OCC1=CC=CC=C1

Specifications

Melting Point 61°C to 65°C
Boiling Point 213°C to 215°C (3.5 mmHg)
Quantity 25 g
Beilstein 1464258
Sensitivity Air Sensitive
Solubility Information Soluble in chloroform,methanol.
Formula Weight 242.27
Percent Purity 98%
Chemical Name or Material 4-Benzyloxy-3-methoxybenzaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.