missing translation for 'onlineSavingsMsg'
Learn More

4'-Benzyloxy-3'-methoxyacetophenone, 98%, Thermo Scientific Chemicals

Product Code. 11318165
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11318165 5 g 5g
11308165 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11318165 Supplier Thermo Scientific Alfa Aesar Supplier No. L08430.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1835-11-6 | Molecular Formula: C16H16O3 | Molecular Weight: 256.301 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1835-11-6
Molecular Formula C16H16O3
Molecular Weight (g/mol) 256.301
MDL Number MFCD00017326
InChI Key HRUAWSQBQLYDKH-UHFFFAOYSA-N
Synonym 1-4-benzyloxy-3-methoxyphenyl ethanone, 4'-benzyloxy-3'-methoxyacetophenone, 4-benzyloxy-3-methoxyacetophenone, acetophenone, 4'-benzyloxy-3'-methoxy, 1-3-methoxy-4-phenylmethoxy phenyl ethanone, 1-4-benzyloxy-3-methoxyphenyl ethan-1-one, 1-3-methoxy-4-phenylmethoxyphenyl ethanone, ethanone, 1-3-methoxy-4-phenylmethoxy phenyl
PubChem CID 99215
IUPAC Name 1-(3-methoxy-4-phenylmethoxyphenyl)ethanone
SMILES CC(=O)C1=CC(=C(C=C1)OCC2=CC=CC=C2)OC

Specifications

Melting Point 85°C to 87°C
Quantity 5 g
Beilstein 1885776
Formula Weight 256.3
Percent Purity 98%
Chemical Name or Material 4'-Benzyloxy-3'-methoxyacetophenone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.