missing translation for 'onlineSavingsMsg'
Learn More

4'-Benzyloxy-3'-methoxyacetophenone, 98%, Thermo Scientific Chemicals

Product Code. 11308165
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11308165 1 g 1g
11318165 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11308165 Supplier Thermo Scientific Alfa Aesar Supplier No. L08430.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 1835-11-6
Molecular Formula C16H16O3
Molecular Weight (g/mol) 256.301
MDL Number MFCD00017326
InChI Key HRUAWSQBQLYDKH-UHFFFAOYSA-N
Synonym 1-4-benzyloxy-3-methoxyphenyl ethanone, 4'-benzyloxy-3'-methoxyacetophenone, 4-benzyloxy-3-methoxyacetophenone, acetophenone, 4'-benzyloxy-3'-methoxy, 1-3-methoxy-4-phenylmethoxy phenyl ethanone, 1-4-benzyloxy-3-methoxyphenyl ethan-1-one, 1-3-methoxy-4-phenylmethoxyphenyl ethanone, ethanone, 1-3-methoxy-4-phenylmethoxy phenyl
PubChem CID 99215
IUPAC Name 1-(3-methoxy-4-phenylmethoxyphenyl)ethanone
SMILES CC(=O)C1=CC(=C(C=C1)OCC2=CC=CC=C2)OC

Especificaciones

Melting Point 85°C to 87°C
Quantity 1 g
Beilstein 1885776
Formula Weight 256.3
Percent Purity 98%
Chemical Name or Material 4'-Benzyloxy-3'-methoxyacetophenone

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.