missing translation for 'onlineSavingsMsg'
Learn More

4-Benzyloxy-3,5-dimethylbenzoic acid, 98+%

Product Code. p-7046485
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11353576 1 g 1g
11363576 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11353576 Supplier Thermo Scientific Alfa Aesar Supplier No. L14536.03
Add to basket
This item is not returnable. View return policy

CAS: 97888-80-7 | C16H16O3 | 256.301 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97888-80-7
Molecular Formula C16H16O3
Molecular Weight (g/mol) 256.301
MDL Number MFCD00272572
InChI Key JABUPJCJZZNUFK-UHFFFAOYSA-N
Synonym 4-benzyloxy-3,5-dimethylbenzoic acid, 4-benzyloxy-3,5-dimethyl-benzoic acid, 3,5-dimethyl-4-phenylmethoxy benzoic acid, 4-benzyloxy-3,5-dimethyl-benzoate
PubChem CID 2733993
IUPAC Name 3,5-dimethyl-4-phenylmethoxybenzoic acid
SMILES CC1=CC(=CC(=C1OCC2=CC=CC=C2)C)C(=O)O

Specifications

Melting Point 157°C to 160°C
Quantity 1 g
Beilstein 5045861
Formula Weight 256.3
Percent Purity ≥98%
Chemical Name or Material 4-Benzyloxy-3,5-dimethylbenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.