missing translation for 'onlineSavingsMsg'
Learn More

4-Benzyloxy-2-fluorobenzeneboronic acid, 96%, Thermo Scientific™

Product Code. 11359385
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11359385 250 mg 250mg
11335035 1 g 1g
11345035 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11359385 Supplier Thermo Scientific Alfa Aesar Supplier No. L18521.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 166744-78-1
Molecular Formula C13H12BFO3
Molecular Weight (g/mol) 246.044
MDL Number MFCD02093064
InChI Key PQWKGFALWCOVTC-UHFFFAOYSA-N
Synonym 4-benzyloxy-2-fluorophenylboronic acid, 4-benzyloxy-2-fluorobenzeneboronic acid, 4-benzyloxy-2-fluorophenyl boronic acid, boronic acid, 2-fluoro-4-phenylmethoxy phenyl, 2-fluoro-4-phenylmethoxyphenyl boronic acid, 4-benzyloxy-2-fluorophenylboronic acid, may contain varying amounts of anhydride, pubchem7760, akos brn-0077, acmc-209dv0
PubChem CID 2737786
IUPAC Name (2-fluoro-4-phenylmethoxyphenyl)boronic acid
SMILES B(C1=C(C=C(C=C1)OCC2=CC=CC=C2)F)(O)O

Specifications

Quantity 250 mg
Formula Weight 246.04
Percent Purity 96%
Chemical Name or Material 4-Benzyloxy-2-fluorobenzeneboronic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.