missing translation for 'onlineSavingsMsg'
Learn More

4-Benzyloxy-2(1H)-pyridone, 98%, Thermo Scientific Chemicals

Product Code. 10587790
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53937-02-3
Molecular Formula C12H11NO2
Molecular Weight (g/mol) 201.23
MDL Number MFCD04039771
InChI Key DOVNUEPFPBWTSV-UHFFFAOYSA-N
Synonym 4-benzyloxy-2-1h-pyridone, 4-benzyloxy-2 1h-pyridone, 4-benzyloxy pyridin-2 1h-one, 4-benzyloxypyridin-2-one, 4-benzyloxy pyridin-2-ol, 4-benzyloxy-2-hydroxypyridine, 4-benzyloxy-1h-pyridin-2-one, 2 1h-pyridinone, 4-phenylmethoxy, 4-benzyloxy-1,2-dihydropyridin-2-one
PubChem CID 3607724
SMILES O=C1NC=CC(OCC2=CC=CC=C2)=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 199.7°C to 201.7°C
Color Brown to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
IUPAC Name 4-phenylmethoxy-1H-pyridin-2-one
Formula Weight 201.23
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 4-Benzyloxy-2(1H)-pyridone
Show More Show Less
compliance-icons
Product Identifier
  • 4-Benzyloxy-2(1H)-pyridone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'