missing translation for 'onlineSavingsMsg'
Learn More

4-Benzyloxy-1-bromo-2-fluorobenzene, 98%

Product Code. 15433857
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 15433857

Brand: Thermo Scientific Alfa Aesar H30053.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 185346-79-6 | C13H10BrFO | 281.124 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 185346-79-6
Molecular Formula C13H10BrFO
Molecular Weight (g/mol) 281.124
MDL Number MFCD09801041
InChI Key BYTJTXKQBSNGCL-UHFFFAOYSA-N
Synonym 4-benzyloxy-1-bromo-2-fluorobenzene, 4-benzyloxy-2-fluorobromobenzene, benzene,1-bromo-2-fluoro-4-phenylmethoxy, acmc-1c1vq, benzyloxy-4-bromo-3-fluorobenzene, 1-benzyloxy-4-bromo-3-fluorobenzene, benzyl 4-bromo-3-fluorophenyl ether, 1-bromo-2-fluoro-4-benzyloxy benzene
PubChem CID 22273933
IUPAC Name 1-bromo-2-fluoro-4-phenylmethoxybenzene
SMILES C1=CC=C(C=C1)COC2=CC(=C(C=C2)Br)F

Specifications

Melting Point 33°C to 35°C
Quantity 5 g
Formula Weight 281.13
Percent Purity 98%
Chemical Name or Material 4-Benzyloxy-1-bromo-2-fluorobenzene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.