missing translation for 'onlineSavingsMsg'
Learn More

4-Benzylmorpholine-2-carboxylic acid hydrochloride, 97%, Thermo Scientific™

Product Code. 15545206 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15545206 1 g 1g
15555206 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15545206 Supplier Thermo Scientific Acros Supplier No. 456690010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 135072-15-0
Molecular Formula C12H15NO3·ClH
Molecular Weight (g/mol) 257.72
InChI Key CEDXMALCJZSQHA-UHFFFAOYSA-N
Synonym 4-benzyl-2-morpholinecarboxylic acid hydrochloride, 4-benzylmorpholine-2-carboxylic acid hydrochloride, 4-benzyl-2-carboxymorpholine hydrochloride, 4-benzyl-2-morpholinecarboxylicacidhydrochloride, 4-benzyl-2-morpholinecarboxylic acid hcl, 4-benzyl-morpholine-2-carboxylic acid hydrochloride, 2-morpholinecarboxylic acid, 4-phenylmethyl-, hydrochloride, 2-morpholinecarboxylicacid, 4-phenylmethyl-, hydrochloride 1:1, acmc-1by3p, ksc173i6l
PubChem CID 2776352
IUPAC Name 4-benzylmorpholine-2-carboxylic acid;hydrochloride
SMILES C1COC(CN1CC2=CC=CC=C2)C(=O)O.Cl

Specifications

Melting Point 244.0°C to 248.0°C
Elemental Analysis 53.69 to 58.16% (C) 5.22 to 5.65% (N)
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass Bottle
Quantity 1 g
Formula Weight 257.72
Percent Purity 97%
Chemical Name or Material 4-Benzylmorpholine-2-carboxylic acid hydrochloride
compliance-icons
Product Identifier
  • 4-Benzylmorpholine-2-carboxylic acid hydrochloride, 98%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.