missing translation for 'onlineSavingsMsg'
Learn More

4-Benzylmorpholine-2-carboxylic acid hydrochloride, 97%, Thermo Scientific™

Product Code. 15499246
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15499246 250 mg 250mg
15479246 1 g 1g
15489246 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15499246 Supplier Thermo Scientific Alfa Aesar Supplier No. H64918.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 135072-15-0
Molecular Formula C12H16ClNO3
Molecular Weight (g/mol) 257.714
MDL Number MFCD02682031
InChI Key CEDXMALCJZSQHA-UHFFFAOYSA-N
Synonym 4-benzyl-2-morpholinecarboxylic acid hydrochloride, 4-benzylmorpholine-2-carboxylic acid hydrochloride, 4-benzyl-2-carboxymorpholine hydrochloride, 4-benzyl-2-morpholinecarboxylicacidhydrochloride, 4-benzyl-2-morpholinecarboxylic acid hcl, 4-benzyl-morpholine-2-carboxylic acid hydrochloride, 2-morpholinecarboxylic acid, 4-phenylmethyl-, hydrochloride, 2-morpholinecarboxylicacid, 4-phenylmethyl-, hydrochloride 1:1, acmc-1by3p, ksc173i6l
PubChem CID 2776352
IUPAC Name 4-benzylmorpholine-2-carboxylic acid;hydrochloride
SMILES C1COC(CN1CC2=CC=CC=C2)C(=O)O.Cl

Specifications

Melting Point 244°C to 245°C
Quantity 250 mg
UN Number UN2811
Beilstein 14403086
Formula Weight 257.71
Percent Purity 97%
Chemical Name or Material 4-Benzylmorpholine-2-carboxylic acid hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.