missing translation for 'onlineSavingsMsg'
Learn More

4-Benzylbiphenyl, 98+%

Product Code. 11448346
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11448346

Brand: Thermo Scientific Alfa Aesar A15392.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 613-42-3 | C19H16 | 244.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 613-42-3
Molecular Formula C19H16
Molecular Weight (g/mol) 244.34
MDL Number MFCD00041831
InChI Key AGPLQTQFIZBOLI-UHFFFAOYSA-N
Synonym 4-benzylbiphenyl, 4-phenylmethyl-1,1'-biphenyl, 1,1'-biphenyl, 4-phenylmethyl, benzylbiphenyl, p-benzylbiphenyl, 4-phenyldiphenylmethane, 1-phenyl-4-benzylbenzene, 1, 4-phenylmethyl, acmc-1bh4u, 1-benzyl-4-phenyl-benzene
PubChem CID 69174
IUPAC Name 1-benzyl-4-phenylbenzene
SMILES C(C1=CC=CC=C1)C1=CC=C(C=C1)C1=CC=CC=C1

Specifications

Melting Point 84°C to 87°C
Boiling Point 285°C to 286°C (110 mmHg)
Flash Point 208°C (406°F)
Quantity 5 g
Beilstein 2047066
Formula Weight 244.34
Percent Purity ≥98%
Chemical Name or Material 4-Benzylbiphenyl

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.