missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Benzylbenzaldehyde, 97%

Product Code. 15477457
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
This item is not returnable. View return policy

Product Code. 15477457

Brand: Alfa Aesar™ H35042.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 67468-65-9
Molecular Formula C14H12O
Molecular Weight (g/mol) 196.25
MDL Number MFCD01666418
InChI Key DHARILAASAAGJJ-UHFFFAOYSA-N
Synonym 4-benzyl-benzaldehyde, p-benzylbenzaldehyde, benzaldehyde, p-benzyl, benzaldehyde, 4-phenylmethyl, ambkkkkk178, 4-phenylmethyl benzaldehyde, 4-07-00-01406 beilstein handbook reference, benzaldehyde,4-phenylmethyl
PubChem CID 49886
IUPAC Name 4-benzylbenzaldehyde
SMILES O=CC1=CC=C(CC2=CC=CC=C2)C=C1

Specifications

Quantity 250 mg
Sensitivity Air sensitive
Formula Weight 196.24
Percent Purity 97%
Chemical Name or Material 4-Benzylbenzaldehyde
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.