Learn More
4-Benzyl-4-hydroxypiperidine, 97%, Thermo Scientific Chemicals
£26.70 - £350.00
Chemical Identifiers
CAS | 51135-96-7 |
---|---|
Molecular Formula | C12H17NO |
Molecular Weight (g/mol) | 191.274 |
MDL Number | MFCD00023147 |
InChI Key | KJZBZOFESQSBCV-UHFFFAOYSA-N |
Synonym | 4-benzyl-4-hydroxypiperidine, 4-piperidinol, 4-benzyl, 4-benzyl-4-piperidinol, 4-piperidinol, 4-phenylmethyl, acmc-20amoy, pubchem16262, 4benzyl-piperidin-4-ol, maybridge3_002978, 4-benzyl-piperidin-4-ol, 4-benzyl-4-piperidinol # |
PubChem CID | 98152 |
IUPAC Name | 4-benzylpiperidin-4-ol |
SMILES | C1CNCCC1(CC2=CC=CC=C2)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11477587
|
Thermo Scientific Alfa Aesar
A15664.03 |
1 g |
£26.70
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11487587
|
Thermo Scientific Alfa Aesar
A15664.06 |
5 g |
£89.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11497587
|
Thermo Scientific Alfa Aesar
A15664.14 |
25 g |
£350.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is a building block used for the synthesis of various compounds, having therapeutic properties, such as synthesis of 4-Benzimidazolyl-piperidinylcarbonyl-piperidine analogs as histamine H3 antagonists.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is a building block used for the synthesis of various compounds, having therapeutic properties, such as synthesis of 4-Benzimidazolyl-piperidinylcarbonyl-piperidine analogs as histamine H3 antagonists.
Solubility
Slightly soluble in water.
Notes
Hygroscopic. Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Chemical Identifiers
51135-96-7 | |
191.274 | |
KJZBZOFESQSBCV-UHFFFAOYSA-N | |
98152 | |
C1CNCCC1(CC2=CC=CC=C2)O |
C12H17NO | |
MFCD00023147 | |
4-benzyl-4-hydroxypiperidine, 4-piperidinol, 4-benzyl, 4-benzyl-4-piperidinol, 4-piperidinol, 4-phenylmethyl, acmc-20amoy, pubchem16262, 4benzyl-piperidin-4-ol, maybridge3_002978, 4-benzyl-piperidin-4-ol, 4-benzyl-4-piperidinol # | |
4-benzylpiperidin-4-ol |
Specifications
51135-96-7 | |
C12H17NO | |
1451803 | |
4-benzyl-4-hydroxypiperidine, 4-piperidinol, 4-benzyl, 4-benzyl-4-piperidinol, 4-piperidinol, 4-phenylmethyl, acmc-20amoy, pubchem16262, 4benzyl-piperidin-4-ol, maybridge3_002978, 4-benzyl-piperidin-4-ol, 4-benzyl-4-piperidinol # | |
KJZBZOFESQSBCV-UHFFFAOYSA-N | |
4-benzylpiperidin-4-ol | |
98152 | |
97% |
∼80°C | |
MFCD00023147 | |
Hygroscopic | |
Slightly soluble in water. | |
C1CNCCC1(CC2=CC=CC=C2)O | |
191.274 | |
191.27 | |
4-Benzyl-4-hydroxypiperidine |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
missing translation for 'einecsNumber' : 257-003-7
missing translation for 'rtecsNumber' : TN6836000
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only