missing translation for 'onlineSavingsMsg'
Learn More

4-Benzyl-2-morpholinecarboxylic acid hydrochloride, 97%, Thermo Scientific™

Product Code. 10252922
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10252922 1 g 1g
10407042 250 mg 250mg
10197814 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10252922 Supplier Thermo Scientific Maybridge Supplier No. CC12401DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 135072-15-0
Molecular Formula C12H16ClNO3
Molecular Weight (g/mol) 257.714
MDL Number MFCD02682031
InChI Key CEDXMALCJZSQHA-UHFFFAOYSA-N
Synonym 4-benzyl-2-morpholinecarboxylic acid hydrochloride, 4-benzylmorpholine-2-carboxylic acid hydrochloride, 4-benzyl-2-carboxymorpholine hydrochloride, 4-benzyl-2-morpholinecarboxylicacidhydrochloride, 4-benzyl-2-morpholinecarboxylic acid hcl, 4-benzyl-morpholine-2-carboxylic acid hydrochloride, 2-morpholinecarboxylic acid, 4-phenylmethyl-, hydrochloride, 2-morpholinecarboxylicacid, 4-phenylmethyl-, hydrochloride 1:1, acmc-1by3p, ksc173i6l
PubChem CID 2776352
IUPAC Name 4-benzylmorpholine-2-carboxylic acid;hydrochloride
SMILES C1COC(CN1CC2=CC=CC=C2)C(=O)O.Cl

Specifications

CAS Min % 97%
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 257.72
Percent Purity 97%
Chemical Name or Material 4-Benzyl-2-morpholinecarboxylic acid hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.