missing translation for 'onlineSavingsMsg'
Learn More

4-Benzyl-2-(chloromethyl)morpholine, 97%, Thermo Scientific™

Product Code. 10416774
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10416774 5 g 5g
10427262 1 g 1g
10437262 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10416774 Supplier Thermo Scientific Maybridge Supplier No. TL01000DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 40987-25-5
Molecular Formula C12H16ClNO
Molecular Weight (g/mol) 225.72
MDL Number MFCD02681886
InChI Key GVWRZZNYCOTWNN-UHFFFAOYNA-N
Synonym 4-benzyl-2-chloromethyl morpholine, 4-benzyl-2-chloromethyl-morpholine, 2-chloromethyl-4-benzylmorpholine, morpholine, 2-chloromethyl-4-phenylmethyl, pubchem12932, 3-chloromethyl-4-phenylmethyl morpholine, 2-chloromethyl-4-benzyl morpholine, 2-chloromethyl-4-benzyl-morpholine
PubChem CID 2801562
IUPAC Name 4-benzyl-2-(chloromethyl)morpholine
SMILES ClCC1CN(CC2=CC=CC=C2)CCO1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Quantity 5 g
Formula Weight 225.72
Percent Purity 97%
Chemical Name or Material 4-Benzyl-2-(chloromethyl)morpholine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.