missing translation for 'onlineSavingsMsg'
Learn More

4-Benzoylpyridine, 98%

Product Code. 10275480 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
Packaging:
Glass bottle
Plastic bottle
Unit Size:
100g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10275480 100 g Plastic bottle 100g
10570332 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10275480 Supplier Thermo Scientific Acros Supplier No. 154091000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 14548-46-0 | C12H9NO | 183.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14548-46-0
Molecular Formula C12H9NO
Molecular Weight (g/mol) 183.21
MDL Number MFCD00006430
InChI Key SKFLCXNDKRUHTA-UHFFFAOYSA-N
Synonym 4-benzoylpyridine, phenyl pyridin-4-yl methanone, phenyl 4-pyridyl ketone, methanone, phenyl-4-pyridinyl, phenyl 4-pyridinyl methanone, 4-pyridyl phenyl ketone, pyridine, 4-benzoyl, ketone, phenyl 4-pyridyl, gamma-benzoylpyridine, 4-benzoyl pyridine
PubChem CID 26731
IUPAC Name phenyl(pyridin-4-yl)methanone
SMILES C1=CC=C(C=C1)C(=O)C2=CC=NC=C2

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 68.0°C to 72.0°C
Color Yellow
Boiling Point 315.0°C
Flash Point 150°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Plastic bottle
Linear Formula C6H5CO(C5H4N)
Quantity 100 g
Beilstein 21, 331
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in methanol,very soluble in ether & ethanol
Formula Weight 183.21
Percent Purity 98%
Physical Form Crystalline Powder and/or Chunks
Chemical Name or Material 4-Benzoylpyridine, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Benzoylpyridine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.