missing translation for 'onlineSavingsMsg'
Learn More

4-Benzoylpiperidine Hydrochloride, TRC

Product Code. 30346295
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30346295 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30346295 Supplier Toronto Research Chemicals Supplier No. TRCB208700100MG
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Benzoylpiperidine, Hydrochloride
CAS 25519-80-6
Molecular Formula C12 H15 N O . Cl H
Quantity 100 mg
Synonym Umeclidinium Bromide Impurity 14 HCl
InChI Formula InChI=1S/C12H15NO.ClH/c14-12(10-4-2-1-3-5-10)11-6-8-13-9-7-11;/h1-5,11,13H,6-9H2;1H
SMILES Cl.O=C(C1CCNCC1)c2ccccc2
IUPAC Name phenyl(piperidin-4-yl)methanone;hydrochloride
Molecular Weight (g/mol) 225.71
Formula Weight 225.092
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.