missing translation for 'onlineSavingsMsg'
Learn More

4-Benzoylbiphenyl, TRC

Product Code. 30008743
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30008743 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30008743 Supplier Toronto Research Chemicals Supplier No. TRCB19822025G
 Restricted Item
Estimated Shipment: 31-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 2128-93-0
Molecular Formula C19 H14 O
Quantity 25 g
Synonym Benzophenone, 4-phenyl- (6CI,7CI,8CI), [1,1'-Biphenyl]-4-ylphenylmethanone, (Biphenyl-4-yl)(phenyl)methanone, 4-(Phenylcarbonyl)biphenyl, 4-Benzoyl-1,1'-biphenyl, 4-Benzoylbiphenyl, 4-Biphenylyl phenyl ketone, 4-Phenylbenzophenone, 4-Phenylphenyl phenyl methanone, Eusolex 3490, Genocure PBZ, NSC 55283, NSC 97365, Phenyl p-biphenylyl ketone, Runtecure 1059, Speedcure BPZ, Speedcure PBZ, Trigonal 12, p-Benzoylbiphenyl, p-Biphenylyl phenyl ketone, p-Phenylbenzophenone
InChI Formula InChI=1S/C19H14O/c20-19(17-9-5-2-6-10-17)18-13-11-16(12-14-18)15-7-3-1-4-8-15/h1-14H
SMILES O=C(c1ccccc1)c2ccc(cc2)c3ccccc3
IUPAC Name phenyl-(4-phenylphenyl)methanone
Molecular Weight (g/mol) 258.31
Formula Weight 258.1045
Chemical Name or Material 4-Phenylbenzophenon
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.