missing translation for 'onlineSavingsMsg'
Learn More

4-(Azidomethyl)benzeneboronic acid pinacol ester, 95%

Product Code. 15494206
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15494206 5 g 5g
15484206 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15494206 Supplier Thermo Scientific Alfa Aesar Supplier No. H58401.06
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1239481-05-0 | C13H18BN3O2 | 259.116 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Notes
Incompatible with oxidizing agents. Store in a dry, cool conditions in a well sealed containers. Store at 4°C.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1239481-05-0
Molecular Formula C13H18BN3O2
Molecular Weight (g/mol) 259.116
MDL Number MFCD20922785
InChI Key CARNXRHOIGMOQD-UHFFFAOYSA-N
Synonym 2-4-azidomethyl phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, amtb503, 4-azidomethyl benzeneboronic acid pinacol ester
PubChem CID 46872199
IUPAC Name 2-[4-(azidomethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)CN=[N+]=[N-]

Specifications

Quantity 5 g
Formula Weight 260.12
Percent Purity 95%
Chemical Name or Material 4-(Azidomethyl)benzeneboronic acid pinacol ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.