missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Azido-L-phenylalanine, MedChemExpress
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
Description
4-Azido-L-phenylalanine is an unnatural amino acid, which is used as an effective vibrational reporter of local protein environments.
Specifications
Specifications
| Chemical Name or Material | 4-Azido-L-phenylalanine |
| Physical Form | Solid |
| CAS | 33173-53-4 |
| Color | Light Nude |
| Quantity | 500 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C9H10N4O2 |
| Synonym | p-Azidophenylalanine p-Azido-L-phenylalanine |
| Solubility Information | H2O : 50 mg/mL (242.48 mM; ultrasonic and adjust pH to 13 with NaOH) ∣H2O : 25 mg/mL (121.24 mM; ultrasonic and adjust pH to 11 with NaOH) ∣DMSO : 5 mg/mL (24.25 mM; ultrasonic and warming and adjust pH to 3 with HCl and heat to 80°C) |
| SMILES | O=C(O)[C@@H](N)CC1=CC=C(N=[N+]=[N-])C=C1 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction