missing translation for 'onlineSavingsMsg'
Learn More

4-Azido-L-phenylalanine hydrochloride, MedChemExpress

Product Code. 16427156
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 16427156

Brand: Medchem Express HY16714A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress 4-Azido-L-phenylalanine hydrochloride is an unnatural amino acid, which is used as an effective vibrational reporter of local protein environments.

4-Azido-L-phenylalanine hydrochloride is an unnatural amino acid, which is used as an effective vibrational reporter of local protein environments.

Specifications

Chemical Name or Material 4-Azido-L-phenylalanine hydrochloride
Physical Form Solid
CAS 34670-43-4
Color Khaki
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C9H11ClN4O2
Synonym p-Azidophenylalanine hydrochloride p-Azido-L-phenylalanine hydrochloride
Solubility Information DMSO : ≥ 103.3 mg/mL (425.70 mM) ∣H2O : 10 mg/mL (41.21 mM; Need ultrasonic)
SMILES O=C(O)[C@@H](N)CC1=CC=C(N=[N+]=[N-])C=C1.Cl
Molecular Weight (g/mol) 242.66
Formula Weight 242.66
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.