missing translation for 'onlineSavingsMsg'
Learn More

4-Aminothiophenol, 97%

Product Code. 11428026
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11428026 100 g 100g
11408026 5 g 5g
11418026 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11428026 Supplier Thermo Scientific Alfa Aesar Supplier No. A14082.22
 Restricted Item
Estimated Shipment: 03-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 1193-02-8 | C6H6NS | 124.18 g/mol

4-Aminothiophenol is used in the synthesis of pectin-4-aminothiophenol conjugates and it forms self-assembled monolayers (SAMs) in the development of DNA. It is used in the detection of thrombin by protein recognition using surface-enhanced Raman spectroscopy (SERS), which finds applications in silver and gold nano particles. It acts as a linker to thiol (SH) functionalizes multiwalled carbon nanotubes (MWCNTs).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Aminothiophenol is used in the synthesis of pectin-4-aminothiophenol conjugates and it forms self-assembled monolayers (SAMs) in the development of DNA. It is used in the detection of thrombin by protein recognition using surface-enhanced Raman spectroscopy (SERS), which finds applications in silver and gold nano particles. It acts as a linker to thiol (SH) functionalizes multiwalled carbon nanotubes (MWCNTs).

Solubility
Immiscible with water.

Notes
Air sensitive. Store in a cool place. Incompatible with acids, acid chlorides, acid anhydrides, chloroformates and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1193-02-8
Molecular Formula C6H6NS
Molecular Weight (g/mol) 124.18
MDL Number MFCD00007880
InChI Key WCDSVWRUXWCYFN-UHFFFAOYSA-M
Synonym 4-aminothiophenol, 4-mercaptoaniline, p-aminothiophenol, p-aminobenzenethiol, 4-amino thiophenol, benzenethiol, 4-amino, p-mercaptoaniline, benzenethiol, p-amino, p-aminophenylmercaptan, 4-aminophenyl mercaptan
PubChem CID 14510
IUPAC Name 4-aminobenzenethiol
SMILES NC1=CC=C([S-])C=C1

Specifications

Melting Point 37°C to 42°C
Boiling Point 156°C to 157°C
Flash Point >110°C (230°F)
Assay Percent Range 97%
Odor Stench
Quantity 100 g
UN Number UN3263
Beilstein 906904
Sensitivity Air Sensitive
Solubility Information Immiscible with water.
Formula Weight 125.19
Percent Purity 97%
Physical Form Crystalline
Chemical Name or Material 4-Aminothiophenol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.