missing translation for 'onlineSavingsMsg'
Learn More

4-(Aminosulfonyl)-N-cyclopropylbenzamide, TRC

Product Code. 30314934
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30314934 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30314934 Supplier Toronto Research Chemicals Supplier No. TRCA453055500MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-(Aminosulfonyl)-N-cyclopropylbenzamide
CAS 1044135-16-1
Molecular Formula C10 H12 N2 O3 S
Quantity 500 mg
Synonym Benzamide, 4-(aminosulfonyl)-N-cyclopropyl-, 4-(Aminosulfonyl)-N-cyclopropylbenzamide, AE 0852999
InChI Formula InChI=1S/C10H12N2O3S/c11-16(14,15)9-5-1-7(2-6-9)10(13)12-8-3-4-8/h1-2,5-6,8H,3-4H2,(H,12,13)(H2,11,14,15)
SMILES NS(=O)(=O)c1ccc(cc1)C(=O)NC2CC2
IUPAC Name N-cyclopropyl-4-sulfamoylbenzamide
Molecular Weight (g/mol) 240.28
Formula Weight 240.0569
Product Title
Select an issue

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.