missing translation for 'onlineSavingsMsg'
Learn More

4-Aminopyridine, 98%

Product Code. 10439010 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
Packaging:
Glass bottle
Unit Size:
100g
1kg
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10439010 100 g Glass bottle 100g
10122610 5 g Glass bottle 5g
10030640 25 g Glass bottle 25g
13458298 1 kg Glass bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10439010 Supplier Thermo Scientific Acros Supplier No. 104571000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 504-24-5 | C5H6N2 | 94.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 504-24-5
Molecular Formula C5H6N2
Molecular Weight (g/mol) 94.12
MDL Number MFCD00006439
InChI Key NUKYPUAOHBNCPY-UHFFFAOYSA-N
Synonym 4-aminopyridine, fampridine, 4-pyridinamine, dalfampridine, 4-pyridylamine, avitrol, p-aminopyridine, 4-ap, ampyra, gamma-aminopyridine
PubChem CID 1727
ChEBI CHEBI:34385
IUPAC Name pyridin-4-amine
SMILES C1=CN=CC=C1N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 157°C to 161°C
Color White to Yellow
Boiling Point 273°C
Flash Point 156°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula (C5H4N)NH2
Quantity 100 g
Beilstein 22,433
Merck Index 15,2802
Solubility Information Solubility in water: 74g/L water (20°C). Other solubilities: soluble in ethanol,benzene and ether,slightly soluble in acetone and methanol
Formula Weight 94.12
Percent Purity 98%
Physical Form Crystalline Powder, Crystals or Solid
Chemical Name or Material 4-Aminopyridine, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Aminopyridine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H311-Toxic in contact with skin.
  • H335-May cause respiratory irritation.
  • H300-Fatal if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.