missing translation for 'onlineSavingsMsg'
Learn More

4-Aminopyridine-3-carboxaldehyde, 95%

Product Code. 11916361 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11916361 1 g 1g
11926361 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11916361 Supplier Thermo Scientific Acros Supplier No. 437350010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 42373-30-8 | C6H7N2O | 123.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42373-30-8
Molecular Formula C6H7N2O
Molecular Weight (g/mol) 123.13
MDL Number MFCD04038972
InChI Key GTPZHMGXKZIHKW-UHFFFAOYSA-O
Synonym 4-aminonicotinaldehyde, 4-amino-3-formylpyridine, 4-amino-pyridine-3-carbaldehyde, 4-aminopyridine-3-carboxaldehyde, 4-amino-3-pyridinecarboxaldehyde, 4-amino-pyridine-3-carboxaldehyde, 3-pyridinecarboxaldehyde, 4-amino, zlchem 837, pubchem7681, acmc-1aqvh
PubChem CID 2761033
IUPAC Name 4-aminopyridine-3-carbaldehyde
SMILES NC1=CC=[NH+]C=C1C=O

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 111.2°C to 112.7°C
Color Cream to White
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 122.13
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 4-Aminopyridine-3-carboxaldehyde
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.