missing translation for 'onlineSavingsMsg'
Learn More

4'-Aminopropiophenone, 98%, Thermo Scientific Chemicals

Product Code. p-4836964 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10416452 10 g 10g
11965321 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10416452 Supplier Thermo Scientific Acros Supplier No. 401040100
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 70-69-9
Molecular Formula C9H11NO
Molecular Weight (g/mol) 149.19
MDL Number MFCD00017113
InChI Key FSWXOANXOQPCFF-UHFFFAOYSA-N
Synonym 4'-aminopropiophenone, p-aminopropiophenone, 1-4-aminophenyl propan-1-one, 1-4-aminophenyl-1-propanone, 1-propanone, 1-4-aminophenyl, 4-aminopropiophenone, propiophenone, 4'-amino, usaf uctl-1856, ethyl p-aminophenyl ketone, paraminopropiophenone
PubChem CID 6270
IUPAC Name 1-(4-aminophenyl)propan-1-one
SMILES CCC(=O)C1=CC=C(N)C=C1

Specifications

Melting Point 135.0°C to 140.0°C
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula NH2C6H4C(O)CH2CH3
Quantity 10 g
Merck Index 15,466
Solubility Information Solubility in water: soluble. Other solubilities: slightly soluble in ethanol, insoluble in diethyl ether
Formula Weight 149.19
Percent Purity 98%
Chemical Name or Material 4'-Aminopropiophenone
compliance-icons
Product Identifier
  • 4'-Aminopropiophenone
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
Precautionary Statement
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.