missing translation for 'onlineSavingsMsg'
Learn More

4-Aminophthalonitrile, 97%

Product Code. 10167183 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
Packaging:
Glass bottle
Unit Size:
10g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10167183 5 g Glass bottle 5g
10661822 10 g Glass bottle 10g
10103061 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10167183 Supplier Thermo Scientific Acros Supplier No. 401020050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 56765-79-8 | C8H5N3 | 143.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56765-79-8
Molecular Formula C8H5N3
Molecular Weight (g/mol) 143.15
MDL Number MFCD00041741
InChI Key RRCAJFYQXKPXOJ-UHFFFAOYSA-N
Synonym 4-aminophthalonitrile, 3,4-dicyanoaniline, 1,2-benzenedicarbonitrile, 4-amino, 3,4-dicyanoanline, 4-amino-1,2-benzenedicarbonitrile, 4-amino-phthalonitril, 4-amino-phthalonitrile, 3,4-dicyano aniline, acmc-1awu3
PubChem CID 92529
IUPAC Name 4-aminobenzene-1,2-dicarbonitrile
SMILES C1=CC(=C(C=C1N)C#N)C#N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 177.0°C to 182.0°C
Color Orange
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Beilstein 05,
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in methanol
Formula Weight 143.15
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 4-Aminophthalonitrile, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Aminophthalonitrile
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
Hazard Statement
  • H311-Toxic in contact with skin.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.