missing translation for 'onlineSavingsMsg'
Learn More

4-Aminophenylether, Dr. Ehrenstorfer

Product Code. 30327111
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30327111 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30327111 Supplier Dr. Ehrenstorfer Supplier No. DREC10215000100MG
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Aminophenylether (4,4'-Oxydianiline)
CAS 101-80-4
Analyte or Component Names 4-Aminophenylether (4,4'-Oxydianiline)
Shipping Condition +20°C
Molecular Formula C12 H12 N2 O
Certifications/Compliance ISO 17034
Synonym 4,4'-Oxybis-benzenamine, Aniline, 4,4'-oxydi- (6CI,8CI), 4,4'-Oxybis[benzenamine], 1-Amino-4-(4-aminophenoxy)benzene, 4,4'-Diaminobiphenyl ether, 4,4'-Diaminobiphenyl oxide, 4,4'-Diaminodiphenyl ether, 4,4'-Diaminodiphenyl oxide, 4,4'-Diaminophenyl ether, 4,4'-Oxybis(aniline), 4,4'-Oxydianiline, 4,4'-Oxydiphenylamine, 4-(4-Aminophenoxy)aniline, 4-(4-Aminophenoxy)phenylamine, 4-(4'-Aminophenoxy)benzenamine, 4-Aminophenyl ether, Bis(4-aminophenyl) ether, Bis(p-aminophenyl) ether, DDE, DPE, DPE (amine), Di(4-aminophenyl) ether, NSC 37075, NSC 6089, ODA, Oxybis(4-aminobenzene), Oxydi-p-phenylenediamine, Oxydianiline, p,p'-Diaminodiphenyl ether, p,p'-Oxybis[aniline], p,p'-Oxydianiline, p-Aminophenyl ether
InChI Formula InChI=1S/C12H12N2O/c13-9-1-5-11(6-2-9)15-12-7-3-10(14)4-8-12/h1-8H,13-14H2
SMILES Nc1ccc(Oc2ccc(N)cc2)cc1
IUPAC Name 4-(4-aminophenoxy)aniline
Molecular Weight (g/mol) 200.24
Quantity 100 mg
Formula Weight 200.095 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.