missing translation for 'onlineSavingsMsg'
Learn More

4-Aminophenylacetonitrile, 99%

Product Code. 11423694
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11423694 100 g 100g
11403694 5 g 5g
11413694 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11423694 Supplier Thermo Scientific Alfa Aesar Supplier No. A11304.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 3544-25-0 | C8H8N2 | 132.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3544-25-0
Molecular Formula C8H8N2
Molecular Weight (g/mol) 132.17
MDL Number MFCD00007912
InChI Key YCWRFIYBUQBHJI-UHFFFAOYSA-N
Synonym 4-aminobenzyl cyanide, 4-aminophenylacetonitrile, 2-4-aminophenyl acetonitrile, 4-aminophenyl acetonitrile, p-aminobenzyl cyanide, benzeneacetonitrile, 4-amino, 4-aminobenzylcyanide, 4-amino-phenyl-acetonitrile, 2-4-aminophenyl ethanenitrile, 4-aminobenzyl
PubChem CID 77000
IUPAC Name 2-(4-aminophenyl)acetonitrile
SMILES NC1=CC=C(CC#N)C=C1

Specifications

Melting Point 42°C to 46°C
Boiling Point 312°C
Flash Point >110°C (230°F)
Quantity 100 g
UN Number UN3439
Beilstein 1100108
Formula Weight 132.17
Percent Purity 99%
Chemical Name or Material 4-Aminophenylacetonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.