missing translation for 'onlineSavingsMsg'
Learn More

4-Aminophenylacetic acid, 98%

Product Code. 11437656
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11437656 10 g 10g
11447656 50 g 50g
11457656 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11437656 Supplier Thermo Scientific Alfa Aesar Supplier No. A14675.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1197-55-3 | C8H9NO2 | 151.17 g/mol

4-Aminophenylacetic acid, is used as a non-translocated competitive inhibitor of the epithelial peptide transporter PepT1. It can be detected using HPLC, NMR techniques.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Aminophenylacetic acid, is used as a non-translocated competitive inhibitor of the epithelial peptide transporter PepT1. It can be detected using HPLC, NMR techniques.

Solubility
Very soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible to strong oxidizing agents.Stable under normal temperatures and pressures. Stable at room temperature in closed containers under normal storage and handling conditions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1197-55-3
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.17
MDL Number MFCD00007916
InChI Key CSEWAUGPAQPMDC-UHFFFAOYSA-N
Synonym 4-aminophenylacetic acid, 2-4-aminophenyl acetic acid, benzeneacetic acid, 4-amino, p-aminophenylacetic acid, 4-aminobenzeneacetic acid, 4-carboxymethylaniline, acetic acid, p-aminophenyl, para-aminophenyl acetic acid, p-aminophenyl acetic acid, 4-aminophenyl acetic acid
PubChem CID 14533
IUPAC Name 2-(4-aminophenyl)acetic acid
SMILES NC1=CC=C(CC(O)=O)C=C1

Specifications

Melting Point ∼201°C (decomposition)
Beilstein 2208094
Merck Index 14,463
Solubility Information Very soluble in water.
Quantity 10 g
Formula Weight 151.17
Percent Purity 98%
Chemical Name or Material 4-Aminophenylacetic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.