missing translation for 'onlineSavingsMsg'
Learn More

4-Aminophenylacetic acid, 98%

Product Code. 10020630 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass Bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10020630 25 g Glass Bottle 25g
10018661 5 g Glass Bottle 5g
10177503 100 g Glass Bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10020630 Supplier Thermo Scientific Acros Supplier No. 104280250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1197-55-3 | C8H9NO2 | 151.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1197-55-3
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.17
MDL Number MFCD00007916
InChI Key CSEWAUGPAQPMDC-UHFFFAOYSA-N
Synonym 4-aminophenylacetic acid, 2-4-aminophenyl acetic acid, benzeneacetic acid, 4-amino, p-aminophenylacetic acid, 4-aminobenzeneacetic acid, 4-carboxymethylaniline, acetic acid, p-aminophenyl, para-aminophenyl acetic acid, p-aminophenyl acetic acid, 4-aminophenyl acetic acid
PubChem CID 14533
IUPAC Name 2-(4-aminophenyl)acetic acid
SMILES NC1=CC=C(CC(O)=O)C=C1

Specifications

Melting Point 196.0°C to 201.0°C
CAS Min % 97.5
CAS Max % 100.0
Color Yellow to Beige
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Linear Formula H2NC6H4CH2CO2H
Beilstein 14, 456
Merck Index 15, 459
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alkalies, soluble in alcohol
Water 1% max. (K.F.)
Quantity 25 g
Formula Weight 151.16
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 4-Aminophenylacetic acid
Show More Show Less
compliance-icons
Product Identifier
  • 4-Aminophenylacetic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.