missing translation for 'onlineSavingsMsg'
Learn More

4-Aminophenyl sulfone, 97%

Product Code. 10102660 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Unit Size:
100g
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10102660 500 g 500g
10192650 5 g 5g
10244320 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10102660 Supplier Thermo Scientific Acros Supplier No. 104365000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 80-08-0 | C12H12N2O2S | 248.3 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80-08-0
Molecular Formula C12H12N2O2S
Molecular Weight (g/mol) 248.3
MDL Number MFCD00007887
InChI Key MQJKPEGWNLWLTK-UHFFFAOYSA-N
Synonym dapsone, diaphenylsulfone, 4,4'-diaminodiphenyl sulfone, dapson, 4,4'-sulfonyldianiline, diaphenylsulfon, sulfona, 4-aminophenyl sulfone, avlosulfone, diphenasone
PubChem CID 2955
ChEBI CHEBI:4325
IUPAC Name 4-(4-aminophenyl)sulfonylaniline
SMILES C1=CC(=CC=C1N)S(=O)(=O)C2=CC=C(C=C2)N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 173°C to 178°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula (H2NC6H4)2SO2
Quantity 500 g
Beilstein 13, 536
Merck Index 15, 2822
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in diluted hydrochloric acid,soluble in alcohol,methanol and acetone
Formula Weight 248.3
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 4-Aminophenyl sulfone, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Aminophenyl sulfone
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.