missing translation for 'onlineSavingsMsg'
Learn More

4-Aminophenol, TRC

Product Code. 30007746
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30007746 50 g 50g
17974065 1 g 1g
17993625 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30007746 Supplier Toronto Research Chemicals Supplier No. TRCA61892050G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 123-30-8
Molecular Formula C6 H7 N O
Molecular Weight (g/mol) 109.13
Synonym Phenol, p-amino- (8CI), 4-Aminophenol, 1-Amino-4-hydroxybenzene, 4-Amino-1-hydroxybenzene, 4-Hydroxy-1-aminobenzene, 4-Hydroxyaniline, 4-Hydroxybenzenamine, 4-Hydroxyphenylamine, A 0384, Activol, Azol, BASF Ursol P Base, Benzofur P, C.I. 76550, C.I. Oxidation Base 6, Certinal, Citol, Durafur Brown RB, Fouramine P, Fourrine 84, Fourrine P Base, Furro P base, NSC 1545, Nako Brown R, Paranol, Pelagol Grey P Base, Pelagol P Base, Renal AC, Rodinal, Tertral P Base, Unal, Ursol P, Ursol P Base, Zoba Brown P Base, p-Aminophenol, p-Hydroxyaniline, p-Hydroxyphenylamine
IUPAC Name 4-aminophenol
SMILES Nc1ccc(O)cc1

Spécification

Analyte or Component Names 4-Aminophenol
Shipping Condition Room Temperature
InChI Formula InChI=1S/C6H7NO/c7-5-1-3-6(8)4-2-5/h1-4,8H,7H2
Solution Type Neat
Industry Type Pharmaceutical
Quantity 50 g
Formula Weight 109.0528
Percent Purity >95%
Chemical Name or Material 4-Aminophenol
Grade HPLC
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.