missing translation for 'onlineSavingsMsg'
Learn More

4-Aminophenol, 98%

Product Code. p-7021300
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
50g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11349429 50 g 50g
11430104 250 g 250g
11420104 1000 g 1000g
11440104 5000 g 5000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11349429 Supplier Thermo Scientific Alfa Aesar Supplier No. A13581.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 123-30-8 | C6H7NO | 109.128 g/mol

4-Aminophenol is used as a dye for textiles, hair, furs and feathers. It is also used as a photographic developer. It finds application as a chemical intermediate in pharmaceuticals and dyes. Further, it acts as a final intermediate in the preparation of paracetamol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Aminophenol is used as a dye for textiles, hair, furs and feathers. It is also used as a photographic developer. It finds application as a chemical intermediate in pharmaceuticals and dyes. Further, it acts as a final intermediate in the preparation of paracetamol.

Solubility
Soluble in acetonitrile, ethyl acetate, acetone, hot water, dimethylsulfoxide and alkalies. Slightly soluble in toluene, diethyl ether, ethanol, trifluoroacetic acid and cold water. Insoluble in benzene and chloroform.

Notes
Incompatible with acids, chloroformates and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-30-8
Molecular Formula C6H7NO
Molecular Weight (g/mol) 109.128
MDL Number MFCD00007869
InChI Key PLIKAWJENQZMHA-UHFFFAOYSA-N
Synonym p-aminophenol, 4-hydroxyaniline, p-hydroxyaniline, phenol, 4-amino, paranol, 4-aminobenzenol, certinal, citol, azol, fouramine p
PubChem CID 403
ChEBI CHEBI:17602
IUPAC Name 4-aminophenol
SMILES C1=CC(=CC=C1N)O

Specifications

Melting Point 186°C to 190°C
Density 1.29
Boiling Point 284°C (decomposition)
Flash Point 195°C (383°F)
Assay Percent Range 98%
Odor Phenol-like
Quantity 50 g
UN Number UN2512
Beilstein 385836
Sensitivity Air and light sensitive
Merck Index 14,462
Solubility Information Soluble in acetonitrile,ethyl acetate,acetone,hot water,dimethylsulfoxide and alkalies. Slightly soluble in toluene,diethyl ether,ethanol,trifluoroacetic acid and cold water. Insoluble in benzene and chloroform.
Formula Weight 109.13
Percent Purity 98%
Chemical Name or Material 4-Aminophenol
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 4-Aminophenol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Germ cell mutagenicity Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H341-Suspected of causing genetic defects.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: 4-Aminophenol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.